Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUDS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022491.2 | NP_071936.2 | 1, 2 | 287 | ||
XM_011538671.2 | XP_011536973.1 | 2, 3 | 1364 | |||
XM_017019818.1 | XP_016875307.1 | 1, 2 | 329 | |||
XM_017019819.1 | XP_016875308.1 | 1, 2 | 288 | |||
XM_017019820.1 | XP_016875309.1 | 1, 2 | 111 | |||
GenBank mRNA | AK024460.1 | - | 1, 2 | 270 | ||
BC098404.1 | - | 1, 2 | 310 | 30332266 | ||
BX647687.1 | - | 2, 3 | 245 |