Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM160B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022749.5 | NP_073586.5 | 8 | 1079 | ||
XM_005273602.3 | XP_005273659.1 | 8 | 1079 | |||
XM_006716381.2 | XP_006716444.1 | 8 | 1079 | |||
XM_006716385.3 | XP_006716448.1 | 9 | 2574 | |||
XM_017013732.1 | XP_016869221.1 | 8 | 1076 | |||
XM_017013733.1 | XP_016869222.1 | 9 | 1196 | |||
XM_017013734.1 | XP_016869223.1 | 8 | 1060 | |||
GenBank mRNA | AF495722.1 | - | 3 | 486 | ||
AK055191.1 | - | 2 | 138 | |||
BC052237.1 | - | 8 | 1076 | |||
BC112147.1 | - | 2 | 168 |