Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301860.1 | NP_001288789.1 | 20 | 2692 | ||
NR_126051.1 | 20 | 2610 | ||||
XM_011528996.1 | XP_011527298.1 | 20 | 2858 | |||
XM_011528997.2 | XP_011527299.1 | 21 | 3064 | |||
XM_011528998.2 | XP_011527300.1 | 20 | 2881 | |||
XM_011528999.2 | XP_011527301.1 | 21 | 3011 | |||
XM_011529000.1 | XP_011527302.1 | 20 | 2855 | |||
XM_011529001.1 | XP_011527303.1 | 19 | 2537 | |||
XM_011529002.2 | XP_011527304.1 | 19 | 2660 | |||
XM_017028020.1 | XP_016883509.1 | 20 | 2687 | |||
GenBank mRNA | BC140806.1 | - | 20 | 2573 | 9021674 | |
BX647082.1 | - | 17 | 3670 | |||
BX648609.1 | - | 1 | 691 |