Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022904.1 | NP_075055.1 | 9 | 1402 | ||
XM_011528185.1 | XP_011526487.1 | 9 | 1402 | |||
XM_011528186.1 | XP_011526488.1 | 9 | 1384 | |||
XM_011528187.1 | XP_011526489.1 | 9 | 1402 | |||
GenBank mRNA | AK024488.1 | - | 8 | 1847 | ||
AK122587.1 | - | 9 | 1397 |