Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321970.1 | NP_001308899.1 | 6 | 766 | ||
NM_001321972.1 | NP_001308901.1 | 6 | 1890 | |||
NM_022839.4 | NP_073750.2 | 6 | 769 | |||
NM_176805.3 | NP_789775.1 | 5 | 670 | |||
NR_135915.1 | 5 | 750 | ||||
NR_135917.1 | 6 | 749 | ||||
NR_135918.1 | 6 | 707 | ||||
GenBank mRNA | AB049944.1 | - | 6 | 769 | ||
AK026165.1 | - | 6 | 514 | |||
AK027059.1 | - | 6 | 1635 | |||
AK222933.1 | - | 6 | 514 | |||
AK223152.1 | - | 6 | 514 | |||
AK315407.1 | - | 6 | 514 | |||
BC009451.1 | - | 5 | 402 | |||
BC012489.1 | - | 6 | 523 | 4452949 | ||
BC032378.1 | - | 6 | 538 | 5110633 | ||
BE621879.1 | - | 1 | 353 | |||
CX163380.1 | - | 2 | 96 |