Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016050.4 | NP_057134.1 | 5 | 598 | ||
NM_170738.3 | NP_733934.1 | 5 | 520 | |||
NM_170739.3 | NP_733935.1 | 6 | 692 | |||
NR_120588.1 | 5 | 590 | ||||
NR_120589.1 | 5 | 448 | ||||
GenBank mRNA | AB049638.1 | - | 5 | 570 | ||
AF151871.1 | - | 5 | 582 | |||
AK127603.1 | - | 5 | 1207 | |||
AK223112.1 | - | 5 | 574 | |||
AK290084.1 | - | 5 | 576 | |||
BC005002.1 | - | 5 | 577 | 3542758 | ||
BC108277.1 | - | 5 | 493 | 4416083 | ||
BQ020382.1 | - | 1 | 258 | |||
BU597133.1 | - | 5 | 602 |