Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193388.3 | NP_001180317.2 | 14 | 1911 | ||
NM_001193389.1 | NP_001180318.1 | 5 | 769 | |||
NM_001276414.1 | NP_001263343.1 | 5 | 965 | |||
XM_005255505.1 | XP_005255562.1 | 14 | 1940 | |||
XM_011522610.1 | XP_011520912.1 | 15 | 2000 | |||
XM_011522611.1 | XP_011520913.1 | 15 | 2000 | |||
XM_011522612.1 | XP_011520914.1 | 14 | 1768 | |||
XM_017023568.1 | XP_016879057.1 | 5 | 766 | |||
XM_017023569.1 | XP_016879058.1 | 5 | 881 | |||
GenBank mRNA | AK022685.1 | - | 3 | 500 | ||
AK027013.1 | - | 5 | 770 | |||
AK124977.1 | - | 4 | 1045 | |||
AM944365.1 | - | 14 | 1873 | |||
BC000150.1 | - | 5 | 927 | 2900118 |