Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023078.3 | NP_075566.2 | 3 | 337 | ||
GenBank mRNA | AK001500.2 | - | 3 | 274 | ||
AK023914.1 | - | 3 | 297 | |||
AK222685.1 | - | 307 | ||||
AK225404.1 | - | 3 | 286 | |||
AK294615.1 | - | 4 | 405 | |||
AK295444.1 | - | 3 | 278 | |||
AK301225.1 | - | 3 | 292 | |||
AL833857.1 | - | 2 | 181 | |||
BC007993.1 | - | 3 | 317 | 3533609 | ||
DA403767.1 | - | 3 | 337 | |||
DA629534.1 | - | 3 | 335 |