Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM173A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271285.1 | NP_001258214.1 | 3 | 520 | ||
NM_023933.2 | NP_076422.1 | 3 | 520 | |||
GenBank mRNA | AK123186.1 | - | 2 | 1242 | ||
BC001181.1 | - | 3 | 504 | 3355322 | ||
BC002624.2 | - | 3 | 472 | 3606040 | ||
BI597021.1 | - | 3 | 525 | |||
BI838160.1 | - | 3 | 503 |