Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDRGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023935.2 | NP_076424.1 | 1 | 111 | ||
GenBank mRNA | AK310809.1 | - | 1 | 94 | ||
BC000643.1 | - | 1 | 63 | 3344932 | ||
BC007957.1 | - | 1 | 69 | 4301923 | ||
BC011851.1 | - | 1 | 64 | 4136696 | ||
HY128886.1 | - | 2 | 258 |