Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321439.1 | NP_001308368.1 | 9 | 1089 | ||
NM_001321440.1 | NP_001308369.1 | 8 | 972 | |||
NM_024029.4 | NP_076934.1 | 9 | 1360 | |||
XM_011528270.2 | XP_011526572.1 | 9 | 1089 | |||
XM_011528271.2 | XP_011526573.1 | 8 | 950 | |||
GenBank mRNA | BC000056.2 | - | 9 | 1008 | 3506385 | |
BC013014.2 | - | 9 | 1029 | 2960233 | ||
BE275116.1 | - | 4 | 452 | |||
BM554546.1 | - | 8 | 891 | |||
BX327495.2 | - | 2 | 227 |