Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321439.1 | NP_001308368.1 | 3 | 277 | ||
NM_001321440.1 | NP_001308369.1 | 3 | 277 | |||
NM_024029.4 | NP_076934.1 | 3 | 548 | |||
XM_011528270.2 | XP_011526572.1 | 3 | 277 | |||
XM_011528271.2 | XP_011526573.1 | 3 | 225 | |||
GenBank mRNA | BC000056.2 | - | 3 | 196 | 3506385 | |
BC013014.2 | - | 3 | 217 | 2960233 | ||
BM554546.1 | - | 3 | 195 | |||
HY020827.1 | - | 3 | 282 |