Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VKORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311311.1 | NP_001298240.1 | 4 | 694 | ||
NM_024006.5 | NP_076869.1 | 3 | 610 | |||
NM_206824.2 | NP_996560.1 | 2 | 500 | |||
GenBank mRNA | AF176924.1 | - | 4 | 634 | ||
AK129513.1 | - | 2 | 298 | |||
AK289790.1 | - | 2 | 399 | |||
AK312005.1 | - | 3 | 523 | |||
AY423044.1 | - | 3 | 340 | |||
AY466113.1 | - | 3 | 552 | |||
AY521634.1 | - | 3 | 340 | |||
BC000828.1 | - | 3 | 340 | |||
BC002911.2 | - | 3 | 378 | 3944530 | ||
BI822140.1 | - | 3 | 566 | |||
CD249837.1 | - | 301 | ||||
CN484751.1 | - | 4 | 421 | |||
DB064454.1 | - | 3 | 511 | |||
KU178762.1 | - | 3 | 340 | |||
KU178763.1 | - | 2 | 230 |