Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129994.1 | NP_001123466.1 | 4 | 430 | ||
NM_001129995.1 | NP_001123467.1 | 4 | 427 | |||
NM_024076.2 | NP_076981.2 | 4 | 430 | |||
XM_011527296.1 | XP_011525598.1 | 4 | 413 | |||
XM_011527297.2 | XP_011525599.1 | 5 | 464 | |||
XM_011527298.2 | XP_011525600.1 | 5 | 499 | |||
XM_017027283.1 | XP_016882772.1 | 5 | 555 | |||
GenBank mRNA | AK027901.1 | - | 4 | 423 | ||
AK096843.1 | - | 3 | 452 | |||
AK225793.1 | - | 4 | 395 | |||
AK291465.1 | - | 4 | 415 | |||
AK294543.1 | - | 5 | 499 | |||
BC001185.1 | - | 4 | 414 | 3502748 | ||
BC009335.2 | - | 4 | 414 | 4128019 | ||
BP339379.1 | - | 5 | 555 | |||
CN301512.1 | - | 4 | 437 |