Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMUB2 SIRNA Assays ›
Gene Symbol:
TMUB2
Gene Name:
transmembrane and ubiquitin like domain containing 2
Gene Aliases:
FP2653
Chromosome Location:
Chr.17: 44186956 - 44191731 on Build GRCh38
UniGene ID:
Hs.181391
Species:
Human
siRNA ID
HSS149100
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMUB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001076674.1
NP_001070142.1
3
309
NM_024107.2
NP_077012.2
2
241
NM_177441.2
NP_803190.2
2
742
XM_011525198.2
XP_011523500.1
2
645
XM_011525200.2
XP_011523502.1
3
448
XM_011525201.2
XP_011523503.1
3
317
XM_011525203.2
XP_011523505.1
4
435
XM_011525204.2
XP_011523506.1
3
374
XM_011525205.2
XP_011523507.1
3
835
XM_011525206.2
XP_011523508.1
4
521
XM_011525207.2
XP_011523509.1
3
453
XM_011525208.2
XP_011523510.1
2
346
XM_011525209.2
XP_011523511.1
2
322
XM_011525210.2
XP_011523512.1
4
522
XM_011525211.2
XP_011523513.1
3
457
XM_017025047.1
XP_016880536.1
3
748
XM_017025048.1
XP_016880537.1
3
418
XM_017025049.1
XP_016880538.1
2
732
XM_017025050.1
XP_016880539.1
2
346
XM_017025051.1
XP_016880540.1
2
264
XM_017025052.1
XP_016880541.1
3
748
XM_017025053.1
XP_016880542.1
4
435
XM_017025054.1
XP_016880543.1
2
732
XM_017025055.1
XP_016880544.1
2
319
XM_017025056.1
XP_016880545.1
3
335
GenBank mRNA
AF289564.1
-
1
48
AF370373.1
-
2
719
AK021759.1
-
2
294
AK096631.1
-
2
2001
AK294198.1
-
4
518
AL533563.3
-
3
304
AL538440.3
-
3
304
AL833899.1
-
2
742
AY007092.1
-
1
288
AY358780.1
-
2
215
BC001792.1
-
2
255
BC004306.1
-
2
177
BC063489.1
-
2
215
5239969
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline