Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024308.3 | NP_077284.2 | 5 | 824 | ||
XM_011525233.2 | XP_011523535.1 | 4 | 564 | |||
GenBank mRNA | AB209247.1 | - | 1 | 3401 | ||
AK026196.1 | - | 5 | 615 | |||
AK096551.1 | - | 1 | 1443 | |||
AK315735.1 | - | 5 | 745 | |||
AY358712.1 | - | 5 | 721 | |||
BC002731.2 | - | 5 | 710 | 3631177 | ||
CR457356.1 | - | 4 | 373 |