Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024313.2 | NP_077289.1 | 5 | 475 | ||
GenBank mRNA | AK057139.1 | - | 5 | 458 | ||
AK294700.1 | - | 5 | 458 | |||
AK316443.1 | - | 5 | 458 | |||
AL160132.1 | - | 5 | 441 | |||
BC002808.1 | - | 5 | 452 | 3634584 | ||
CR456463.1 | - | 5 | 435 |