Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024337.3 | NP_077313.3 | 1 | 312 | ||
XM_017009845.1 | XP_016865334.1 | 1 | 511 | |||
GenBank mRNA | AY335938.1 | - | 1 | 312 | ||
BC029160.1 | - | 1 | 14 | |||
U90307.1 | - | 1 | 12 |