Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C17ORF62 SIRNA Assays ›
Gene Symbol:
C17orf62
Gene Name:
chromosome 17 open reading frame 62
Gene Aliases:
-
Chromosome Location:
Chr.17: 82442586 - 82450831 on Build GRCh38
UniGene ID:
Hs.163113
Species:
Human
siRNA ID
HSS149182
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C17orf62
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001033046.3
NP_001028218.1
6, 7
614
NM_001100407.2
NP_001093877.1
7, 8
664
NM_001100408.2
NP_001093878.1
5, 6
572
NM_001193653.1
NP_001180582.1
7, 8
884
NM_001193654.1
NP_001180583.1
6, 7
834
NM_001193655.1
NP_001180584.1
6, 7
778
NM_001193657.1
NP_001180586.1
7, 8
831
NR_036514.1
5, 6
540
NR_036516.1
5, 6
491
NR_036517.1
4, 5
449
NR_036518.1
3, 4
375
XM_006722293.2
XP_006722356.1
5, 6
1339
XM_011523606.2
XP_011521908.1
5, 6
1519
XM_017025072.1
XP_016880561.1
5, 6
1519
XM_017025073.1
XP_016880562.1
4, 5
1477
XM_017025074.1
XP_016880563.1
6, 7
612
XM_017025075.1
XP_016880564.1
6, 7
1381
XM_017025076.1
XP_016880565.1
7, 8
609
XM_017025077.1
XP_016880566.1
6, 7
773
XM_017025078.1
XP_016880567.1
5, 6
570
GenBank mRNA
AF269290.1
-
1, 2
742
AK027743.1
-
3, 4
315
AK054876.1
-
5, 6
570
AK056342.1
-
5, 6
447
AK074950.1
-
7, 8
761
AK090484.1
-
5, 6
2685
AK092865.1
-
3, 4
1673
AK225389.1
-
6, 7
520
BC003595.1
-
6, 7
498
2822930
BC004171.2
-
6, 7
498
2822930
BG716477.1
-
6, 7
714
BM147030.1
-
5, 6
396
BM801484.1
-
7, 8
597
CN260096.1
-
5, 6
398
DA008508.1
-
4, 5
346
DA342707.1
-
5, 6
562
DA500882.1
-
5, 6
496
DB113745.1
-
5, 6
395
Back To Top
More Information
Panther Classification:
Molecular Function -
chaperone
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0