Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253815.1 | NP_001240744.1 | 3 | 602 | ||
NM_001253816.1 | NP_001240745.1 | 3 | 582 | |||
NM_024531.4 | NP_078807.1 | 3 | 696 | |||
NR_045600.1 | 3 | 910 | ||||
XM_006716658.2 | XP_006716721.1 | 3 | 1013 | |||
XM_006716659.2 | XP_006716722.1 | 3 | 828 | |||
XM_006716660.2 | XP_006716723.1 | 3 | 979 | |||
XM_011517300.2 | XP_011515602.1 | 2 | 373 | |||
XM_017013819.1 | XP_016869308.1 | 3 | 808 | |||
XM_017013820.1 | XP_016869309.1 | 3 | 543 | |||
GenBank mRNA | AB522904.1 | - | 2 | 485 | ||
AK021918.1 | - | 3 | 866 | |||
AK027888.1 | - | 3 | 575 | |||
AK291581.1 | - | 3 | 602 | |||
AK303822.1 | - | 3 | 501 | |||
AL560856.3 | - | 3 | 721 | |||
AY070774.1 | - | 2 | 418 | |||
BC002917.2 | - | 3 | 648 | 3954480 | ||
CN260061.1 | - | 1 | 264 |
Set Membership: |
G-protein coupled receptor |