Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195302.1 | NP_001182231.1 | 3 | 453 | ||
NM_001204911.1 | NP_001191840.1 | 3 | 453 | |||
NM_024598.3 | NP_078874.2 | 3 | 453 | |||
XM_005256144.3 | XP_005256201.1 | 3 | 328 | |||
XM_011523328.2 | XP_011521630.1 | 3 | 429 | |||
XM_011523329.2 | XP_011521631.1 | 3 | 311 | |||
XM_011523330.1 | XP_011521632.1 | 3 | 419 | |||
GenBank mRNA | AK023216.1 | - | 3 | 425 | ||
AK293220.1 | - | 3 | 263 | |||
AK301494.1 | - | 3 | 403 | |||
AK303121.1 | - | 3 | 408 | |||
BC004415.1 | - | 3 | 389 | 3641317 | ||
BC006291.2 | - | 3 | 379 | 4025993 | ||
BC007774.2 | - | 3 | 383 | 4302289 | ||
BC010099.2 | - | 3 | 424 | |||
BC021554.1 | - | 3 | 373 | 4541229 | ||
DC363309.1 | - | 3 | 454 | |||
DC380401.1 | - | 3 | 453 |