Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195302.1 | NP_001182231.1 | 4, 5 | 663 | ||
NM_024598.3 | NP_078874.2 | 5, 6 | 717 | |||
XM_005256144.3 | XP_005256201.1 | 5, 6 | 592 | |||
XM_011523328.2 | XP_011521630.1 | 5, 6 | 693 | |||
XM_011523329.2 | XP_011521631.1 | 5, 6 | 575 | |||
GenBank mRNA | AK023216.1 | - | 5, 6 | 689 | ||
AK126223.1 | - | 1, 2 | 2324 | |||
AK293220.1 | - | 5, 6 | 527 | |||
AK298473.1 | - | 6, 7 | 564 | |||
AK303121.1 | - | 4, 5 | 618 | |||
BC004415.1 | - | 5, 6 | 653 | 3641317 | ||
BC006291.2 | - | 5, 6 | 643 | 4025993 | ||
BC007774.2 | - | 5, 6 | 647 | 4302289 | ||
BC021554.1 | - | 5, 6 | 637 | 4541229 |