Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282143.1 | NP_001269072.1 | 6 | 1150 | ||
NM_001282144.1 | NP_001269073.1 | 6 | 1558 | |||
NM_001282358.1 | NP_001269287.1 | 6 | 1082 | |||
NM_024618.3 | NP_078894.2 | 6 | 1177 | |||
XM_005271669.1 | XP_005271726.1 | 6 | 1401 | |||
XM_005271670.1 | XP_005271727.1 | 5 | 1054 | |||
XM_005271671.1 | XP_005271728.1 | 5 | 1228 | |||
XM_005271672.1 | XP_005271729.1 | 5 | 1051 | |||
XM_006718904.1 | XP_006718967.1 | 6 | 1435 | |||
XM_011542980.1 | XP_011541282.1 | 6 | 1065 | |||
XM_017018294.1 | XP_016873783.1 | 6 | 1435 | |||
GenBank mRNA | AB094095.1 | - | 6 | 1161 | ||
AK056454.1 | - | 5 | 875 | |||
AK095247.1 | - | 4 | 362 | |||
AK291716.1 | - | 6 | 1144 | |||
AK303002.1 | - | 5 | 1051 | |||
AY245437.1 | - | 5 | 920 | |||
BC013199.1 | - | 2 | 281 | |||
BC110890.1 | - | 6 | 1523 | 4937963 | ||
BX647705.1 | - | 6 | 1135 | |||
EF452237.1 | - | 5 | 920 |