Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282143.1 | NP_001269072.1 | 6 | 1487 | ||
NM_001282144.1 | NP_001269073.1 | 6 | 1895 | |||
NM_001282358.1 | NP_001269287.1 | 6 | 1419 | |||
NM_024618.3 | NP_078894.2 | 6 | 1514 | |||
XM_005271669.1 | XP_005271726.1 | 6 | 1738 | |||
XM_005271670.1 | XP_005271727.1 | 5 | 1391 | |||
XM_005271671.1 | XP_005271728.1 | 5 | 1565 | |||
XM_005271672.1 | XP_005271729.1 | 5 | 1388 | |||
XM_006718904.1 | XP_006718967.1 | 6 | 1772 | |||
XM_011542980.1 | XP_011541282.1 | 6 | 1402 | |||
XM_017018294.1 | XP_016873783.1 | 6 | 1772 | |||
GenBank mRNA | AB094095.1 | - | 6 | 1498 | ||
AK025131.1 | - | 1 | 141 | |||
AK056454.1 | - | 5 | 1212 | |||
AK095247.1 | - | 4 | 699 | |||
AK225223.1 | - | 1 | 141 | |||
AK291716.1 | - | 6 | 1481 | |||
AK303002.1 | - | 5 | 1388 | |||
AY245437.1 | - | 5 | 1257 | |||
BC013199.1 | - | 2 | 618 | |||
BC110890.1 | - | 6 | 1860 | 4937963 | ||
BX647705.1 | - | 6 | 1472 | |||
EF452237.1 | - | 5 | 1257 |