Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YRDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024640.3 | NP_078916.3 | 1, 2 | 393 | ||
GenBank mRNA | AF250397.1 | - | 1, 2 | 212 | ||
AL524812.3 | - | 1, 2 | 427 | |||
AY172561.1 | - | 1, 2 | 385 | |||
AY286019.1 | - | 1, 2 | 374 | |||
AY286020.1 | - | 1, 2 | 374 | |||
BC008984.1 | - | 1, 2 | 420 | |||
BC067857.1 | - | 1, 2 | 378 | |||
BC068057.1 | - | 1, 2 | 355 | |||
BC080186.1 | - | 1, 2 | 395 | 6147134 |