Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324171.1 | NP_001311100.1 | 8, 9 | 776 | ||
NM_001324172.1 | NP_001311101.1 | 8, 9 | 776 | |||
NM_030581.3 | NP_085058.3 | 8, 9 | 749 | |||
XM_005256146.3 | XP_005256203.1 | 8, 9 | 753 | |||
XM_011523331.2 | XP_011521633.1 | 7, 8 | 593 | |||
XM_011523332.2 | XP_011521634.1 | 2, 3 | 176 | |||
XM_017023667.1 | XP_016879156.1 | 7, 8 | 610 | |||
XM_017023668.1 | XP_016879157.1 | 6, 7 | 386 | |||
XM_017023669.1 | XP_016879158.1 | 7, 8 | 481 | |||
XM_017023670.1 | XP_016879159.1 | 6, 7 | 489 | |||
GenBank mRNA | AB067510.1 | - | 3, 4 | 222 | ||
AK302557.1 | - | 8, 9 | 817 | |||
AL562571.3 | - | 6, 7 | 791 | |||
BC004519.2 | - | 8, 9 | 713 | 3938206 | ||
BC014887.2 | - | 2, 3 | 141 |