Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191060.1 | NP_001177989.1 | 3, 4 | 696 | ||
NM_001191061.1 | NP_001177990.1 | 3, 4 | 365 | |||
NM_024698.5 | NP_078974.1 | 3, 4 | 533 | |||
XM_011520369.1 | XP_011518671.1 | 3, 4 | 407 | |||
XM_011520370.1 | XP_011518672.1 | 3, 4 | 475 | |||
XM_011520371.1 | XP_011518673.1 | 3, 4 | 360 | |||
GenBank mRNA | AJ428202.1 | - | 2, 3 | 253 | ||
AK023106.1 | - | 2, 3 | 253 | |||
AK290481.1 | - | 3, 4 | 696 | |||
AK311038.1 | - | 3, 4 | 364 | |||
BC019033.2 | - | 3, 4 | 331 | 4577143 | ||
BC023545.2 | - | 3, 4 | 491 | 4110556 |