Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 632 | |||
NR_136193.1 | 4 | 1329 | ||||
NR_136194.1 | 4 | 799 | ||||
NR_136197.1 | 4 | 703 | ||||
NR_136198.1 | 3 | 520 | ||||
NR_136199.1 | 5 | 842 | ||||
NR_136200.1 | 4 | 1332 | ||||
NR_136201.1 | 4 | 1499 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1240 | ||
AK021601.1 | - | 1 | 979 | |||
AK300846.1 | - | 4 | 710 | |||
BC101437.1 | - | 2 | 352 | |||
BC101981.1 | - | 2 | 352 | |||
BG519698.1 | - | 3 | 398 | |||
BQ882603.1 | - | 4 | 632 | |||
EU000853.1 | - | 4 | 545 | |||
EU000854.1 | - | 4 | 545 | |||
EU000855.1 | - | 3 | 1637 | |||
EU000856.1 | - | 3 | 1637 |