Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 682 | |||
NR_136193.1 | 4 | 1379 | ||||
NR_136194.1 | 4 | 849 | ||||
NR_136197.1 | 4 | 753 | ||||
NR_136198.1 | 3 | 570 | ||||
NR_136199.1 | 5 | 892 | ||||
NR_136200.1 | 4 | 1382 | ||||
NR_136201.1 | 4 | 1549 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1290 | ||
AK021601.1 | - | 1 | 1029 | |||
AK300846.1 | - | 4 | 760 | |||
BC101437.1 | - | 2 | 402 | |||
BC101981.1 | - | 2 | 402 | |||
BG519698.1 | - | 3 | 448 | |||
BQ882603.1 | - | 4 | 682 | |||
EU000853.1 | - | 4 | 595 | |||
EU000854.1 | - | 4 | 595 | |||
EU000855.1 | - | 3 | 1687 | |||
EU000856.1 | - | 3 | 1687 |