Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003679.2 | 3 | 395 | |||
NR_136192.1 | 3 | 389 | ||||
NR_136193.1 | 3 | 386 | ||||
NR_136194.1 | 3 | 556 | ||||
NR_136195.1 | 3 | 389 | ||||
NR_136197.1 | 3 | 460 | ||||
NR_136198.1 | 2 | 277 | ||||
NR_136199.1 | 4 | 599 | ||||
NR_136200.1 | 3 | 389 | ||||
NR_136201.1 | 3 | 556 | ||||
GenBank mRNA | AB075501.1 | - | 3 | 297 | ||
AK054605.1 | - | 3 | 298 | |||
AK123668.1 | - | 3 | 1452 | |||
AK300846.1 | - | 3 | 467 | |||
AU253586.1 | - | 3 | 298 | |||
BC101437.1 | - | 1 | 109 | |||
BC101981.1 | - | 1 | 109 | |||
BG519698.1 | - | 2 | 156 | |||
BQ882603.1 | - | 3 | 389 | |||
DA665500.1 | - | 3 | 465 | |||
DA863153.1 | - | 2 | 263 | |||
DC386781.1 | - | 4 | 510 | |||
DC421261.1 | - | 3 | 460 | |||
EF529495.1 | - | 3 | 395 | |||
EF529496.1 | - | 3 | 395 | |||
EF529497.1 | - | 3 | 395 | |||
EF529498.1 | - | 3 | 395 | |||
EF529499.1 | - | 3 | 302 | |||
EF529500.1 | - | 3 | 395 | |||
EF529501.1 | - | 2 | 190 | |||
EF529502.1 | - | 2 | 190 | |||
EF529503.1 | - | 3 | 469 | |||
EF529504.1 | - | 4 | 511 | |||
EF529505.1 | - | 3 | 395 | |||
EF529506.1 | - | 3 | 395 | |||
EF529507.1 | - | 3 | 395 | |||
EF529508.1 | - | 3 | 395 | |||
EF529509.1 | - | 3 | 395 | |||
EU000853.1 | - | 3 | 302 | |||
EU000854.1 | - | 3 | 302 | |||
EU000855.1 | - | 3 | 302 | |||
EU000856.1 | - | 3 | 302 |