Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010000.2 | NP_001010000.1 | 10 | 1195 | ||
XM_005258144.1 | XP_005258201.2 | 11 | 1517 | |||
XM_005258145.1 | XP_005258202.2 | 11 | 1517 | |||
XM_005258146.2 | XP_005258203.1 | 10 | 1439 | |||
GenBank mRNA | AB037735.1 | - | 11 | 4468 | ||
AK001174.1 | - | 5 | 524 | |||
AK289791.1 | - | 4 | 455 | |||
AK302026.1 | - | 10 | 1104 | |||
AK308259.1 | - | 10 | 1143 | |||
BC033668.1 | - | 4 | 414 | |||
BC065274.1 | - | 9 | 956 | 6164396 | ||
BX648684.1 | - | 10 | 1153 |