Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024768.2 | NP_079044.2 | 5, 6 | 1451 | ||
XM_005247781.3 | XP_005247838.1 | 5, 6 | 1479 | |||
XM_011513161.2 | XP_011511463.1 | 5, 6 | 911 | |||
XM_011513164.2 | XP_011511466.1 | 4, 5 | 1524 | |||
GenBank mRNA | AK022119.1 | - | 3, 4 | 725 | ||
BC093800.1 | - | 1, 2 | 162 | 7939645 | ||
BC112195.1 | - | 1, 2 | 162 | 8327663 |