Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC102B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093729.1 | NP_001087198.1 | 9 | 1502 | ||
NM_024781.2 | NP_079057.2 | 7 | 1396 | |||
XM_011526174.2 | XP_011524476.1 | 6 | 1294 | |||
XM_017025973.1 | XP_016881462.1 | 9 | 1535 | |||
XM_017025974.1 | XP_016881463.1 | 7 | 1538 | |||
XM_017025975.1 | XP_016881464.1 | 10 | 1586 | |||
XM_017025976.1 | XP_016881465.1 | 9 | 1535 | |||
GenBank mRNA | AK027247.1 | - | 5 | 685 | ||
AK225597.1 | - | 5 | 685 | |||
AL833863.1 | - | 6 | 714 | |||
BC126448.1 | - | 5 | 674 | 8992164 | ||
BC126450.1 | - | 5 | 674 | 8992166 | ||
CK823962.1 | - | 2 | 221 | |||
CR749520.1 | - | 7 | 1345 |