Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136040.2 | NP_001129512.1 | 6 | 708 | ||
NM_001142565.1 | NP_001136037.1 | 6 | 681 | |||
NM_024811.3 | NP_079087.3 | 6 | 784 | |||
XM_005274298.4 | XP_005274355.1 | 6 | 767 | |||
XM_005274299.4 | XP_005274356.1 | 6 | 711 | |||
XM_005274303.4 | XP_005274360.1 | 7 | 739 | |||
XM_011545257.2 | XP_011543559.1 | 6 | 794 | |||
XM_011545258.2 | XP_011543560.1 | 6 | 665 | |||
XM_011545259.2 | XP_011543561.1 | 6 | 638 | |||
XM_011545260.2 | XP_011543562.1 | 7 | 785 | |||
XM_011545261.2 | XP_011543563.1 | 7 | 786 | |||
XM_011545262.2 | XP_011543564.1 | 7 | 732 | |||
XM_011545263.2 | XP_011543565.1 | 7 | 731 | |||
XM_017018345.1 | XP_016873834.1 | 6 | 767 | |||
GenBank mRNA | AJ275970.1 | - | 5 | 574 | ||
AK092037.1 | - | 6 | 688 | |||
AK096343.1 | - | 6 | 664 | |||
AK294578.1 | - | 6 | 637 | |||
AL832546.1 | - | 4 | 379 | |||
AL832667.1 | - | 6 | 672 | |||
AL833332.1 | - | 6 | 637 | |||
BC018135.1 | - | 6 | 674 | 3913173 | ||
BX537888.1 | - | 6 | 662 |