Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAQR6 SIRNA Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
PRdelta
Chromosome Location:
Chr.1: 156243321 - 156248488 on Build GRCh38
UniGene ID:
Hs.235873
Species:
Human
siRNA ID
HSS149351
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAQR6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272104.1
NP_001259033.1
4
573
NM_001272105.1
NP_001259034.1
5
645
NM_001272106.1
NP_001259035.1
2
590
NM_001272107.1
NP_001259036.1
3
708
NM_001272109.1
NP_001259038.1
2
590
NM_001272110.1
NP_001259039.1
4
573
NM_001272111.1
NP_001259040.1
4
531
NM_001272112.1
NP_001259041.1
3
455
NM_024897.3
NP_079173.2
3
708
NM_198406.2
NP_940798.1
4
531
NR_073610.1
3
455
XM_005245494.3
XP_005245551.1
3
826
XM_006711546.2
XP_006711609.1
4
471
XM_006711547.3
XP_006711610.1
4
944
XM_006711548.3
XP_006711611.1
3
826
XM_011510000.2
XP_011508302.1
4
902
XM_017002385.1
XP_016857874.1
3
2102
GenBank mRNA
AF455045.1
-
3
627
AF455046.1
-
2
534
AF455047.1
-
3
369
AK054682.1
-
4
512
AK094346.1
-
4
551
AK095143.1
-
4
432
AK095197.1
-
3
643
AK294143.1
-
3
383
AK295915.1
-
3
397
AK295916.1
-
2
546
AK307441.1
-
2
235
AK316034.1
-
5
584
AL834227.1
-
3
599
AY424284.1
-
4
452
BC058509.1
-
4
415
5770270
BC071623.1
-
1
74
BI551776.1
-
4
475
BI819919.1
-
3
375
DT218672.1
-
5
586
FJ200490.1
-
1
27
FJ200491.1
-
1
27
FJ200492.1
-
1
27
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
steroid hormone mediated signaling pathway
response to steroid hormone
steroid hormone receptor activity
steroid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline