Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002451.2 | 3 | 430 | |||
GenBank mRNA | AK024359.1 | - | 3 | 350 | ||
BC098116.1 | - | 4 | 338 | |||
BC098259.1 | - | 4 | 338 | |||
BC105084.1 | - | 2 | 156 | |||
BC112069.1 | - | 2 | 156 | |||
BC143677.1 | - | 3 | 222 | |||
DA501191.1 | - | 3 | 430 |