Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024980.4 | NP_079256.4 | 4 | 1102 | ||
XM_005263496.4 | XP_005263553.1 | 4 | 1012 | |||
XM_005263497.4 | XP_005263554.1 | 3 | 856 | |||
GenBank mRNA | AK292888.1 | - | 4 | 1056 | ||
AK304154.1 | - | 3 | 861 | |||
AY768808.1 | - | 4 | 1102 |
Set Membership: |
G-protein coupled receptor |