Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256599.1 | NP_001243528.1 | 1 | 225 | ||
NM_001256600.1 | NP_001243529.1 | 1 | 225 | |||
NM_001282378.1 | NP_001269307.1 | 1 | 225 | |||
NM_001282379.1 | NP_001269308.1 | 1 | 225 | |||
NM_025058.4 | NP_079334.3 | 1 | 225 | |||
NR_046327.1 | 1 | 225 | ||||
NR_104150.1 | 1 | 225 | ||||
XM_011510002.1 | XP_011508304.1 | 1 | 81 | |||
XM_017002393.1 | XP_016857882.1 | 1 | 298 | |||
GenBank mRNA | AK026882.1 | - | 1 | 136 | ||
AK096158.1 | - | 1 | 103 | |||
AK225075.1 | - | 1 | 136 | |||
AK296300.1 | - | 1 | 221 | |||
AK297318.1 | - | 1 | 133 | |||
AY251386.1 | - | 1 | 45 | |||
BC069416.1 | - | 1 | 121 | 7262300 | ||
BC126372.1 | - | 1 | 142 | 8992088 | ||
BC143828.1 | - | 1 | 142 | |||
DA509424.1 | - | 1 | 225 |