Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025108.2 | NP_079384.2 | 10 | 1319 | ||
XM_011522667.1 | XP_011520969.1 | 9 | 1223 | |||
GenBank mRNA | AK023971.1 | - | 9 | 1400 | ||
AK093788.1 | - | 8 | 1849 | |||
AK128408.1 | - | 7 | 2441 | |||
AK301929.1 | - | 10 | 1315 | |||
BC008882.2 | - | 6 | 1148 | |||
BC018719.1 | - | 10 | 1307 | 4868952 |