Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032548.3 | NP_115937.1 | 11 | 1218 | ||
NM_172027.2 | NP_742024.1 | 11 | 1207 | |||
NR_033429.1 | 11 | 1172 | ||||
XM_006713769.3 | XP_006713832.1 | 11 | 1241 | |||
XM_011513213.1 | XP_011511515.1 | 11 | 1141 | |||
XM_017007285.1 | XP_016862774.1 | 10 | 1070 | |||
XM_017007286.1 | XP_016862775.1 | 10 | 1052 | |||
XM_017007287.1 | XP_016862776.1 | 11 | 1178 | |||
XM_017007288.1 | XP_016862777.1 | 10 | 1111 | |||
GenBank mRNA | AB053324.1 | - | 11 | 1197 | ||
AB053325.1 | - | 11 | 1186 | |||
AB053326.1 | - | 11 | 1151 | |||
AF297986.1 | - | 11 | 1164 | |||
AF447886.1 | - | 10 | 1311 | |||
AK090509.1 | - | 1 | 1579 | |||
AK098738.1 | - | 4 | 338 | |||
AK130594.1 | - | 7 | 697 | |||
AK315148.1 | - | 11 | 1187 | |||
BC011858.2 | - | 11 | 1205 | 4309664 |