Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317089.1 | NP_001304018.1 | 2 | 226 | ||
NM_025218.3 | NP_079494.1 | 4 | 766 | |||
NR_133659.1 | 4 | 2114 | ||||
GenBank mRNA | AB052907.1 | - | 4 | 690 | ||
AF304377.1 | - | 4 | 690 | |||
AF346416.1 | - | 4 | 690 | |||
AF346595.1 | - | 4 | 660 | |||
AF425265.1 | - | 4 | 677 | |||
AK292519.1 | - | 4 | 766 | |||
AK316582.1 | - | 4 | 755 | |||
BC035416.1 | - | 4 | 733 | 4822311 | ||
DB462306.1 | - | 2 | 191 | |||
KT591165.1 | - | 4 | 2081 |