Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316964.1 | NP_001303893.1 | 3 | 588 | ||
NM_001316965.1 | NP_001303894.1 | 3 | 588 | |||
NM_025232.3 | NP_079508.2 | 3 | 588 | |||
GenBank mRNA | AF218014.1 | - | 3 | 366 | ||
AK025899.1 | - | 3 | 518 | |||
AK025930.1 | - | 3 | 221 | |||
AK172846.1 | - | 3 | 523 | |||
AK302352.1 | - | 3 | 405 | |||
AY562242.1 | - | 3 | 119 | |||
BC013048.1 | - | 3 | 572 | 3863838 | ||
BC050622.1 | - | 3 | 490 | 6504298 | ||
BG695963.1 | - | 3 | 177 | |||
CR457353.1 | - | 3 | 119 |