Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030570.3 | NP_085047.1 | 3, 4 | 977 | ||
NM_182683.2 | NP_872624.1 | 4, 5 | 812 | |||
NM_182684.2 | NP_872625.1 | 5, 6 | 892 | |||
GenBank mRNA | AK056760.1 | - | 4, 5 | 620 | ||
AK290096.1 | - | 3, 4 | 880 | |||
AY233462.1 | - | 5, 6 | 725 | |||
BC004304.1 | - | 3, 4 | 801 | 3625247 |
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250612.3 | XP_005250669.1 | 3, 4 | 1298 | ||
XM_006716140.3 | XP_006716203.1 | 5, 6 | 970 | |||
XM_006716141.3 | XP_006716204.1 | 4, 5 | 890 |