Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322462.1 | NP_001309391.1 | 7 | 1733 | ||
NM_001322463.1 | NP_001309392.1 | 5 | 1108 | |||
NM_001322464.1 | NP_001309393.1 | 5 | 1392 | |||
NM_030645.1 | NP_085148.1 | 5 | 1062 | |||
XM_017002404.1 | XP_016857893.1 | 4 | 1265 | |||
XM_017002405.1 | XP_016857894.1 | 4 | 1044 | |||
XM_017002406.1 | XP_016857895.1 | 6 | 1018 | |||
XM_017002407.1 | XP_016857896.1 | 4 | 950 | |||
GenBank mRNA | AB051507.1 | - | 4 | 769 | ||
AK056345.1 | - | 2 | 247 | |||
AK056382.1 | - | 5 | 1062 | |||
AK125138.1 | - | 5 | 1739 | |||
AK125837.1 | - | 7 | 2002 | |||
AK298698.1 | - | 4 | 416 | |||
AK299713.1 | - | 1 | 426 | |||
AL136569.1 | - | 4 | 649 | |||
AY358453.1 | - | 5 | 984 | |||
BC010871.1 | - | 2 | 271 | |||
BC017254.2 | - | 4 | 551 | 3051344 |