Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322462.1 | NP_001309391.1 | 7, 8 | 1766 | ||
NM_001322463.1 | NP_001309392.1 | 5, 6 | 1141 | |||
NM_030645.1 | NP_085148.1 | 5, 6 | 1095 | |||
XM_017002404.1 | XP_016857893.1 | 4, 5 | 1298 | |||
XM_017002405.1 | XP_016857894.1 | 4, 5 | 1077 | |||
XM_017002406.1 | XP_016857895.1 | 6, 7 | 1051 | |||
XM_017002407.1 | XP_016857896.1 | 4, 5 | 983 | |||
GenBank mRNA | AB051507.1 | - | 4, 5 | 802 | ||
AK056345.1 | - | 2, 3 | 280 | |||
AK056382.1 | - | 5, 6 | 1095 | |||
AK125138.1 | - | 5, 6 | 1772 | |||
AK125837.1 | - | 7, 8 | 2035 | |||
AK298698.1 | - | 4, 5 | 449 | |||
AK299713.1 | - | 1, 2 | 459 | |||
AL136569.1 | - | 4, 5 | 682 | |||
AY358453.1 | - | 5, 6 | 1017 | |||
BC010871.1 | - | 2, 3 | 304 | |||
BC017254.2 | - | 4, 5 | 584 | 3051344 |