Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030772.4 | NP_110399.2 | 2 | 1461 | ||
GenBank mRNA | AF271261.1 | - | 1 | 1424 | ||
AK312811.1 | - | 2 | 1422 |
Gene Symbol: GJA9-MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037633.1 | 2 | 1461 | |||
NR_037634.1 | 2 | 1461 | ||||
GenBank mRNA | AK123051.1 | - | 2 | 1455 | ||
BC051675.1 | - | 2 | 1432 |