Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030940.3 | NP_112202.2 | 4 | 1195 | ||
GenBank mRNA | AF284752.1 | - | 4 | 1150 | ||
BC071621.1 | - | 4 | 1129 | 5107151 |