Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030978.2 | NP_112240.1 | 5 | 1574 | ||
XM_011519067.1 | XP_011517369.1 | 3 | 708 | |||
GenBank mRNA | AF087842.1 | - | 3 | 362 | ||
AK056174.1 | - | 5 | 1562 | |||
AK315604.1 | - | 3 | 430 | |||
BC000018.1 | - | 3 | 364 | 3509794 | ||
BC000798.1 | - | 3 | 396 | 3451934 | ||
BC002418.1 | - | 3 | 364 | 3163950 | ||
BX537659.1 | - | 3 | 3843 |