Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042353.2 | NP_001035812.1 | 2 | 568 | ||
NM_001289145.1 | NP_001276074.1 | 3 | 677 | |||
NM_001289146.1 | NP_001276075.1 | 2 | 565 | |||
NM_001289147.1 | NP_001276076.1 | 2 | 355 | |||
NM_031424.5 | NP_113612.1 | 1 | 413 | |||
NM_207121.4 | NP_997004.1 | 2 | 360 | |||
XM_011529381.2 | XP_011527683.1 | 2 | 1101 | |||
GenBank mRNA | AB196290.1 | - | 1 | 235 | ||
AK314165.1 | - | 2 | 549 | |||
BC004222.2 | - | 2 | 360 | 3532660 | ||
BC012800.2 | - | 1 | 376 | 2961135 | ||
BC022238.1 | - | 1 | 362 | 4181303 | ||
BI909945.1 | - | 2 | 483 | |||
BI911808.1 | - | 2 | 526 | |||
BI916595.1 | - | 1 | 422 | |||
CN425669.1 | - | 1 | 329 | |||
CR997106.1 | - | 1 | 165 | |||
DQ431181.1 | - | 1 | 216 |