Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM110A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042353.2 | NP_001035812.1 | 2 | 676 | ||
NM_001289145.1 | NP_001276074.1 | 3 | 785 | |||
NM_001289146.1 | NP_001276075.1 | 2 | 673 | |||
NM_001289147.1 | NP_001276076.1 | 2 | 463 | |||
NM_031424.5 | NP_113612.1 | 1 | 521 | |||
NM_207121.4 | NP_997004.1 | 2 | 468 | |||
XM_011529381.2 | XP_011527683.1 | 2 | 1209 | |||
GenBank mRNA | AB196290.1 | - | 1 | 343 | ||
AK314165.1 | - | 2 | 657 | |||
AL530841.3 | - | 1 | 486 | |||
BC004222.2 | - | 2 | 468 | 3532660 | ||
BC012800.2 | - | 1 | 484 | 2961135 | ||
BC022238.1 | - | 1 | 470 | 4181303 | ||
BI909945.1 | - | 2 | 591 | |||
BI911808.1 | - | 2 | 634 | |||
BI916595.1 | - | 1 | 531 | |||
CR997106.1 | - | 1 | 273 | |||
DQ431181.1 | - | 1 | 324 |